Learn More
Medchemexpress LLC HY-N6722 1mg Medchemexpress, Swainsonine CAS:72741-87-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN67221MG
Encompass_Preferred
Medchemexpress, HY-N6722 1mg Swainsonine CAS:72741-87-8 Swainsonine is an alkaloid isolated from Astragalus, acts as an inhibitor of α-mannosidase, with anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0461