Learn More
Cincinnati Surgical Swann Morton™ Surgical Handle
SDP
Catalog No. 19200283
Change view
Click to view available options
Size:
# 4
Catalog No. Size
19-200-283 # 4
1 options
This item is not returnable. View return policy
Catalog No. 19-200-283 Supplier Cincinnati Surgical Supplier No. 074S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Short handle, size 4
  • Stainless Steel
  • Non-Sterile
  • Fits Blades 20 through 26
  • Specifications
    Blade Material Stainless Steel
    Size # 4
    Length (English) 5.16 in.
    Length (Metric) 131 mm
    For Use With (Equipment) Blade # 20 to # 26
    Disposable Reusable
    Sterility Non-sterile