Learn More
StatLab Swann Morton Surgical Scalpel Blades
SDP
Catalog No. 21033469
Change view
Click to view available options
Size:
# 22
Catalog No. Size
21-033-469 # 22
1 options
Catalog No. 21-033-469 Supplier StatLab Supplier No. SWM22
Add to Cart
Add to Cart
Stainless steel blades, features a curved cutting edge and a flat, unsharpened back edge
  • Features a curved cutting edge and a flat, unsharpened back edge
  • Sterile and Individually Wrapped
  • Blades are packaged 100 per box
Specifications
Product Type Scalpel Blade
Size # 22