Learn More
SWAN ANALYTICAL USA SWANSENSOR REFERENCE NA,12MM
SDP
Catalog No. NC2112261
Encompass
Swansensor Reference Na, Ø 12 mm, screw plug, without cable, incl. 100 ml filling solution
This item is not returnable. View return policy
Catalog No. NC2112261 Supplier SWAN ANALYTICAL USA Supplier No. A87.840.100
Swansensor Reference Na, Ø 12 mm, screw plug, without cable, incl. 100 ml filling solution