Learn More
Pickering Laboratories Inc Artificial Perspiration, ANSI-BHMA A156.189, Not Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501354823
Encompass_Preferred
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. This artificial perspiration formulation is used to test finishes on various base materials as per the builder’s hardware association. This formulation is not offered in a stabilized format.
Catalog No. 50-135-4823 Supplier Pickering Laboratories Inc Supplier No. 17000504
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. This artificial perspiration formulation is used to test finishes on various base materials as per the builder’s hardware association. This formulation is not offered in a stabilized format.