Learn More
ELITechGroup Sweat Controls, L1, L2, L3, 12 ea, 36x0.75mL
SDP
Catalog No. NC9069181
Encompass
Each Sweat control has a different molarity. Sweat Control 1 is approximately 40mmol/L, Sweat Control 2 is approximately 70mmol/L, and Sweat Control 3 is approximately 130mmol/L.
Catalog No. NC9069181 Supplier ELITechGroup Supplier No. SS150
Each Sweat control has a different molarity. Sweat Control 1 is approximately 40mmol/L, Sweat Control 2 is approximately 70mmol/L, and Sweat Control 3 is approximately 130mmol/L.