Learn More
Pickering Laboratories Inc Artificial Eccrine Perspiration-Sebum Emulsion, Not Stabilized (250 mL/bottle)
SDP
Catalog No. 501354806
Encompass_Preferred
Inspired by the ASTM D4265-98 method for staining, the emulsion uses 5% Artificial Sebum in Eccrine Persepiration. Oleic acid and triethanolamine are added as emulsifiers. Prepared without dust/dirt for a more universal application, this emulsion mimics non-exercise induced skin surface film (SSFL). As SSFL, it can be used to test any topical use product or the stability of any article that will come in contact with human sweat. This product requires refrigeration to remain in solution and prevent rancidity.
Catalog No. 50-135-4806 Supplier Pickering Laboratories Inc Supplier No. 17000547
Inspired by the ASTM D4265-98 method for staining, the emulsion uses 5% Artificial Sebum in Eccrine Persepiration. Oleic acid and triethanolamine are added as emulsifiers. Prepared without dust/dirt for a more universal application, this emulsion mimics non-exercise induced skin surface film (SSFL). As SSFL, it can be used to test any topical use product or the stability of any article that will come in contact with human sweat. This product requires refrigeration to remain in solution and prevent rancidity.