Learn More
Cayman Chemical Swerosde 50mg
SDP
Catalog No. 502949643
Encompass_Preferred
An iridoid glycoside; inhibits LPS-induced production of PGE2 and ROS in mouse RAW 264.7 macrophages at 40 µM; inhibits the growth of U937 lymphoma and HL-60 leukemia cells (IC50s = 67.92 and 62.58 µM, respectively); reduces tumor growth in an HL-60 mouse xenograft model at 50 mg/kg; reduces weight gain and serum total cholesterol, triglyceride, LDL-C, and FFA levels in a mouse model of obesity induced by a HFD at 120 mg/kg
Catalog No. 50-294-9643 Supplier Cayman Chemical Supplier No. 3345450 MG
An iridoid glycoside; inhibits LPS-induced production of PGE2 and ROS in mouse RAW 264.7 macrophages at 40 µM; inhibits the growth of U937 lymphoma and HL-60 leukemia cells (IC50s = 67.92 and 62.58 µM, respectively); reduces tumor growth in an HL-60 mouse xenograft model at 50 mg/kg; reduces weight gain and serum total cholesterol, triglyceride, LDL-C, and FFA levels in a mouse model of obesity induced by a HFD at 120 mg/kg