Learn More
Cayman Chemical Swertisin, Purity: ≥98%, CAS: 6991-10-2, 10 mg
SDP
Catalog No. 502910716
Encompass_Preferred
A flavonoid C-glycoside with diverse biological activities; inhibits SGLT2 in HEK293 at 7.5 µg/ml; is an adenosine A1 receptor antagonist (IC50 = 137 µM); inhibits HBV replication in HepG2 2.2.15 cells at 0.2-5 µM; decreases blood glucose levels in a mouse model of streptozotocin-induced diabetes; prevents scopolamine-induced increases in escape latency in the Morris water maze in mice
Catalog No. 50-291-0716 Supplier Cayman Chemical Supplier No. 3474410 MG
A flavonoid C-glycoside with diverse biological activities; inhibits SGLT2 in HEK293 at 7.5 µg/ml; is an adenosine A1 receptor antagonist (IC50 = 137 µM); inhibits HBV replication in HepG2 2.2.15 cells at 0.2-5 µM; decreases blood glucose levels in a mouse model of streptozotocin-induced diabetes; prevents scopolamine-induced increases in escape latency in the Morris water maze in mice