Learn More
Cayman Chemical Swertiamarin, Purity: ≥98%, CAS: 17388-39-5, 100 mg
SDP
Catalog No. 502939389
Encompass_Preferred
A secoiridoid glycoside with diverse biological activities; scavenges ABTS radicals and hydrogen peroxide (IC50s = 2.83 and 5.7 μM, respectively); inhibits NO production in IL-1β-stimulated FLSs isolated from rat hindpaw at 50 μg/ml; inhibits oleate-induced lipid droplet and triglyceride accumulation in HepG2 cells at 25 μg/ml; decreases liver lipid accumulation, ballooning degeneration, and TNF-α and IL-6 levels in a mouse model of fructose-induced NAFLD at 50 mg/kg per day; decreases fasting blood glucose and serum cholesterol levels in rat model of STZ-induced diabetes at 50 mg/kg per day
Catalog No. 50-293-9389 Supplier Cayman Chemical Supplier No. 27634100 MG
A secoiridoid glycoside with diverse biological activities; scavenges ABTS radicals and hydrogen peroxide (IC50s = 2.83 and 5.7 μM, respectively); inhibits NO production in IL-1β-stimulated FLSs isolated from rat hindpaw at 50 μg/ml; inhibits oleate-induced lipid droplet and triglyceride accumulation in HepG2 cells at 25 μg/ml; decreases liver lipid accumulation, ballooning degeneration, and TNF-α and IL-6 levels in a mouse model of fructose-induced NAFLD at 50 mg/kg per day; decreases fasting blood glucose and serum cholesterol levels in rat model of STZ-induced diabetes at 50 mg/kg per day