Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Swertiamarin | 17388-39-5 | MFCD07783984 | 98.0% | 374.34 g·mol⁻¹ | C16H22O10 | 10 MG
SDP

Catalog No. 5000295780
Encompass_Preferred

Swertiamarin is an iridoid glycoside (C16H22O10, MW 374.34 g·mol⁻¹) reported to have hypoglycemic, hypolipidemic, anti-rheumatic, and antioxidant activities. Supplied as an analytical standard for laboratory research, it is used for qualitative and quantitative analyses by HPLC, GC, and MS and is intended for research use only.

  • Suitable for qualitative and quantitative analysis by HPLC, GC, and MS.
  • Reported orally active iridoid with hypoglycemic and antioxidant activity.
  • Provided in small solid quantities appropriate for analytical workflows.
  • Molecular weight 374.34 g·mol⁻¹; chemical formula C16H22O10.
  • Purity commonly specified as 98% by HPLC; see Certificate of Analysis for details.

Catalog No. 50-002-95780 Supplier Medchemexpress LLC Supplier No. HYN080710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Swertiamarin is an iridoid glycoside (C16H22O10, MW 374.34 g·mol⁻¹) reported to have hypoglycemic, hypolipidemic, anti-rheumatic, and antioxidant activities. Supplied as an analytical standard for laboratory research, it is used for qualitative and quantitative analyses by HPLC, GC, and MS and is intended for research use only.

  • Suitable for qualitative and quantitative analysis by HPLC, GC, and MS.
  • Reported orally active iridoid with hypoglycemic and antioxidant activity.
  • Provided in small solid quantities appropriate for analytical workflows.
  • Molecular weight 374.34 g·mol⁻¹; chemical formula C16H22O10.
  • Purity commonly specified as 98% by HPLC; see Certificate of Analysis for details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.