Learn More
Medchemexpress LLC HY-N0807 5mg Medchemexpress, Swertiamarin CAS:17388-39-5 Purity:>98%
SDP
Catalog No. 501970311
Encompass_Preferred
Medchemexpress, HY-N0807 5mg Swertiamarin CAS:17388-39-5 Swertiamarin, a secoiridoid glycoside found in genera of Enicostemma Species, confers anti-hyperglycemic and anti-hyperlipidemic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0311 Supplier Medchemexpress LLC Supplier No. HYN08075MG
Medchemexpress, HY-N0807 5mg Swertiamarin CAS:17388-39-5 Swertiamarin, a secoiridoid glycoside found in genera of Enicostemma Species, confers anti-hyperglycemic and anti-hyperlipidemic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.