Learn More
Motic™ Swift Line Inverted Microscope
SCI_ED
Catalog No. S89537 Shop All Motic Swift Line Products
Change view
Click to view available options
Head Style:
Trinocular
Catalog No. Head Style
S89537 Trinocular
1 options
This item is not returnable. View return policy
Catalog No. S89537 Supplier Motic™ Supplier No. MAE31R
This item is not returnable. View return policy
With a wide range of features for university and laboratory use, this advanced research microscope features infinity corrected and phase contrast optics, LED illumination, and a trinocular head, as well as an extra-long working distance (ELWD) condenser.
Complimenting the illumination system is the extra-long working distance (ELWD) 0.3 N.A. condenser and its 72 mm working distance for larger sample sizes.
This microscope includes a built-in, internal 110/220 V auto-switching power supply. Ergonomically designed, professional frame with low-position coaxial focusing allows one-handed operation, making extended use comfortable.
  • Trinocular, Siedentopf with light splitter (100% or 20/80%)
  • Ergonomic 45 degree observation angle
  • Side-facing quintuple nosepiece
  • Infinity corrected optics: 4X Infinity, 10X and 20X Infinity and Phase
  • LED illumination with intensity control
  • Plain stage with glass and metal inserts
Specifications
Includes IEC Power Cord, Centering Telescope, Phase Slider, Glass and Metal Stage Inserts, Blue and Green Interference Filters, 2 Allen Hex Keys, Dust Cover
Condenser Type ELWD N.A. 0.3 Condenser
Magnification Power 4X, 10X, 20X
Objective Types Infinity and Infinity Phase
Eyepiece WF 10X
Lighting Type LED
Head Style Trinocular
Product Line MAE-31R Series
Product Type Inverted Microscope
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov