Learn More
Swift™ M3-F Forensic Comparison Microscope
SCI_ED
Catalog No. s01897 Shop All Motic Swift Line Products
Change view
Click to view available options
4X, 10XD, 40XRD, micro, 2X macro objectives
Catalog No.
S01897 4X, 10XD, 40XRD, micro, 2X macro objectives
1 options
Catalog No. S01897 Supplier Swift™ Supplier No. M3F
Add to Cart
Add to Cart
Revolutionary technology allows you to compare images in both micro and macro environments.
Powerful optical system allows images to be seen either 100% from the left microscope, 100% from the right, side-by-side or overlapping. The dedicated macro lens, with a large working distance, allows you to view not only bullets but also larger, more bulky “evidence” items.
  • Trinocular head
  • Energy-efficient cordless top and bottom LED illumination
  • Co-axial focusing
  • Gliding stage plate with “one-touch” stage clips
  • Balanced tripod footing
  • Eyepiece: W10XD, 18mm
  • Objectives: 4X, 10XD, 40XRD, micro and 1X macro
Order Info
With the optional C-mount adapter, any C-mount-ready imaging device can be attached, including Motic™ cameras, for analysis with the Motic software provided. (Motic software included with the Swift Cam; eyepiece cameras sold separately.)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov