Learn More
Swift™ Video C Mount Camera Adapter
SCI_ED
Catalog No. S04524 Shop All Motic Swift Line Products
Change view
Click to view available options
For Use With (Equipment):
M3-F, M10T-S, M10T-P, M15T-P Microscopes
Catalog No. For Use With (Equipment)
S04524 M3-F, M10T-S, M10T-P, M15T-P Microscopes
1 options
Catalog No. S04524 Supplier Swift™ Supplier No. MA15602
Add to Cart
Add to Cart
For use with 0.5 in. chip
  • Optional accessory that allows user to easily attach any C-mount ready imaging device, including Moticam cameras, to a Swift microscope eyepiece
  • For use with Swift M3-F, Swift M10T-S, Swift M10T-P and Swift M15T-P model microscopes
Specifications
Type Adapter for M3-F, M10T-S, M10T-P and M15T-P model microscope
For Use With (Equipment) M3-F, M10T-S, M10T-P, M15T-P Microscopes
Head Style C mount
Includes Moticam cameras to eyepiece
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov