Learn More
Bolle Safety™ Swift OTG Safety Spectacles
Catalog No. 17467677
Change view
Click to view available options
Lens Tint:
Clear
Smoke
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
17-467-677 Smoke
17-467-678 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-467-677 Supplier Bolle Safety™ Supplier No. SWIOTN20U
Only null left
Add to Cart
Add to Cart
Safety glasses with anti-fog and anti-scratch features to ensure that your vision remains clear and unobstructed.
Swift Safety Spectacles with standard polycarbonate frame.
  • Scratch-resistant, anti-fog.
  • Reusable.
  • With UV protection.
Specifications
Lens Coating Scratch-resistant, Anti-fog
Lens Tint Smoke
Product Type Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
Frame Type Standard