Learn More
Applied Biosystems™ SwiftArrayStudio™ Microarray Analyzer
Catalog No. 000471
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
00-047-1 1 Ea.
1 options
Catalog No. 00-047-1 Supplier Applied Biosystems™ Supplier No. 000471
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The SwiftArrayStudio Microarray Analyzer is an automated array processing instrument that uniquely integrates hybridization, washing/staining, and scanning functions into a single platform for comprehensive genome-wide single nucleotide polymorphism (SNP) genotyping and copy number variation (CNV) analysis in both human and agricultural genomics applications.
The SwiftArrayStudio Microarray Analyzer is an automated array processing instrument that integrates hybridization, washing/staining, and scanning functions into a single platform for comprehensive genome-wide single nucleotide polymorphism (SNP) genotyping and copy number variation (CNV) analysis in both human and agricultural genomics applications.
As a fully automated array processing instrument, the SwiftArrayStudio analyzer is engineered for high-speed throughput and enhanced reliability, requiring as few as 21 hours of instrument processing time. When utilized in conjunction with the SwiftArray Assay, this integrated system enables researchers to obtain genomic data in as little as 30 hours, representing a significant advancement in automated microarray analysis workflow efficiency.
Features of the SwiftArrayStudio Microarray Analyzer include:
  • High-speed performance—engineered for rapid data processing and analysis to meet demanding computational requirements in research and laboratory environments
  • Automated array processing—integrated automated processing capabilities for efficient handling of multi-dimensional data arrays, reducing manual intervention and processing errors
  • Enhanced throughput—robust system architecture enables flexible throughput, supporting workflows ranging from a single plate in as little as 30 hours to 16 plates processed within a five-day work week
Ultra-high-speed scanning capabilities enable rapid sample processing
The SwiftArrayStudio Microarray Analyzer enables exceptional scanning performance with the ability to process a single sample in as little as 15 seconds. This remarkable speed is achieved through advanced LED illumination technology coupled with a high-resolution 4.1-megapixel imaging system. For complete plate processing, the instrument demonstrates consistent performance across different array densities, requiring as little as 105 minutes for 96-array plates and 95 minutes for 384-array plates. When integrated with the SwiftArray Assay protocol, laboratories can obtain complete results in as little as 30 hours, representing a significant advancement in microarray workflow efficiency that enables accelerated turnaround times and improved overall laboratory productivity.
Fully integrated automation
The SwiftArrayStudio Microarray Analyzer is a fully automated microarray system available that combines hybridization, wash/stain, and scanning operations within a single integrated platform. This comprehensive end-to-end automation architecture enables consistent and reliable results while minimizing the need for manual intervention during processing cycles. A complete single plate can be processed without any manual steps in approximately 21 hours, allowing laboratory personnel to focus their expertise on other critical analytical tasks while helping maintain optimal workflow continuity and reduce the potential for human error in sample processing.
Scalable throughput architecture accommodates variable laboratory demands
The instrument supports flexible throughput, enabling the processing of a single plate in as little as 30 hours up to16 plates processed within a five-day work week. This scalable architecture helps ensure optimal instrument utilization across different workflow scenarios while maintaining consistent processing efficiency. The system's design allows laboratories to transition between low-volume research applications and high-throughput clinical or industrial testing requirements without compromising analytical performance or operational reliability.
Advanced remote diagnostics and support system operational uptime
The SwiftArrayStudio analyzer incorporates sophisticated Smart Help and Remote Support technologies that work synergistically to offer enhanced service experiences and reduce instrument downtime. Smart Help functionality allows users to initiate support tickets directly from the instrument interface, automatically transmitting error logs and comprehensive diagnostic data to help accelerate troubleshooting processes. The Remote Support capability enables service professionals to securely access real-time instrument health data, perform remote diagnostics, and provide guided issue resolution without requiring on-site visits. This integrated support architecture significantly improves service response speed, diagnostic accuracy, and overall system reliability, helping ensure continuous operational availability for critical laboratory workflows.
Intuitive operation through advanced user interface design
The instrument features a touchscreen computer interface with guided user operation that helps simplify complex microarray processing protocols. The streamlined workflow requires users to simply load samples and array plates before initiating the automated processing sequence, helping make the sophisticated analytical capabilities accessible to laboratory personnel with varying levels of technical expertise. This user-centric design philosophy enables consistent operation while reducing training requirements and the potential for operational errors.  
Warranty and Services
1 Year
TRUSTED_SUSTAINABILITY
Specifications
Type SwiftArrayStudio Instrument
Dimensions W x H x D: 54.52 x 25.03 x 26.92 in. (138.48 x 63.58 x 68.39 cm)
For Use With (Application) Microarray Processing
Weight 170.1 kg
Quantity 1 Ea.
For Research Use Only.