Learn More
Sino Biological SwiftFluo TR-FRET 96-Well Low-Volume White Microplates (PS, Solid, Non-treated, No lid) 40 Plates
Catalog No. 5000696255
Encompass_Preferred
SwiftFluo TR-FRET 96-Well Low-Volume White Microplates are specially designed for TR-FRET, Chemiluminescence, Alphaand Isotope Test. The shallow well structure is perfectly adapted to micro-reaction systems of less than 40 µL. The unique 96-well Low-Volume design effectively balances the number of samples, detection volume and signal strength, bringing the liquid surface closer to the detection probe to enhance signal acquisition efficiency. While ensuring detection sensitivity, it significantly reduces reagent dosage. This microplate is manufactured with precision molding technology to ensure well-to-well uniformity and mechanical strength, meeting the requirements of high-throughput screening. It is an ideal laboratory consumables for micro-detection applications, significantly reducing experimental costs and enhancing operational convenience.
Catalog No. 50-006-96255 Supplier Sino Biological Supplier No. SFP0240
SwiftFluo TR-FRET 96-Well Low-Volume White Microplates are specially designed for TR-FRET, Chemiluminescence, Alphaand Isotope Test. The shallow well structure is perfectly adapted to micro-reaction systems of less than 40 µL. The unique 96-well Low-Volume design effectively balances the number of samples, detection volume and signal strength, bringing the liquid surface closer to the detection probe to enhance signal acquisition efficiency. While ensuring detection sensitivity, it significantly reduces reagent dosage. This microplate is manufactured with precision molding technology to ensure well-to-well uniformity and mechanical strength, meeting the requirements of high-throughput screening. It is an ideal laboratory consumables for micro-detection applications, significantly reducing experimental costs and enhancing operational convenience.