Learn More
Sino Biological SwiftFluo TR-FRET BTKs substrate - biotin 250 nmol
Catalog No. 5000696203
Encompass_Preferred
SwiftFluo TR-FRET BTKs substrate - biotin is designed for measuring the Tyrosine kinase (TKs) activity with SwiftFluor™ TR-FRET TKs kinase kit (Cat. # SFKA001)
Catalog No. 50-006-96203 Supplier Sino Biological Supplier No. SFKA015250
SwiftFluo TR-FRET BTKs substrate - biotin is designed for measuring the Tyrosine kinase (TKs) activity with SwiftFluor™ TR-FRET TKs kinase kit (Cat. # SFKA001)