Learn More
Sino Biological SwiftFluo TR-FRET JAKs substrate - biotin 25 nmol
Catalog No. 5000696280
Encompass_Preferred
SwiftFluo TR-FRET JAKs substrate - biotin is designed for measuring the Tyrosine kinase (TKs) activity with SwiftFluor™ TR-FRET TKs kinase kit (Cat. # SFKA001).
Catalog No. 50-006-96280 Supplier Sino Biological Supplier No. SFKA01625
SwiftFluo TR-FRET JAKs substrate - biotin is designed for measuring the Tyrosine kinase (TKs) activity with SwiftFluor™ TR-FRET TKs kinase kit (Cat. # SFKA001).