Learn More
Kingfisher Biotech Inc Swine CCL17 Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Catalog No. 502964660
Encompass
The Swine CCL17 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine CCL17 applications are for cell culture, ELISA Standard, ELISpot Control, and Western Blot Control. Swine CCL17 yeast-derived recombinant protein can be purchased in multiple sizes. Swine CCL17 Specifications: (Molecular Weight:9.0 kDa) (Amino Acid Sequence:ARGTNVGREC CLEYFKGAIP LRKLVGWYQT SDECTRDAIV LVTFQGRAIC SDPKDTWVKK AIRYLKKNMK PQDPVTQKS). For research use only. Made in the USA.
Catalog No. 50-296-4660 Supplier Kingfisher Biotech Inc Supplier No. RP2306S025
The Swine CCL17 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine CCL17 applications are for cell culture, ELISA Standard, ELISpot Control, and Western Blot Control. Swine CCL17 yeast-derived recombinant protein can be purchased in multiple sizes. Swine CCL17 Specifications: (Molecular Weight:9.0 kDa) (Amino Acid Sequence:ARGTNVGREC CLEYFKGAIP LRKLVGWYQT SDECTRDAIV LVTFQGRAIC SDPKDTWVKK AIRYLKKNMK PQDPVTQKS). For research use only. Made in the USA.