Learn More
Kingfisher Biotech Inc Ovinie CCL5 ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY2060S003
Encompass_Preferred
The Ovine CCL5 (RANTES) ELISA contains capture antibody, recombinant protein standard, and detection antibody for development of a Ovine CCL5 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP, C-C motif chemokine ligand 5. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5665