Learn More
Kingfisher Biotech Inc Swine CCL8 Polyclonal Antibody (100 ug)
SDP
Catalog No. 502892013
Encompass
The Swine CCL8 polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Biotech Swine CCL8 ELISA. The Swine CCL8 antibody was produced in rabbits and is antigen-affinity purified. The reactivity by species is: (Swine CCL8 - Strong) (Bat CCL8 - None) (Bovine CCL8 - Moderate) (Canine CCL8 - None) (Cynomolgus Monkey CCL8 - None) (Equine CCL8 - None) (Feline CCL8 - None) (Ferret CCL8 - None) (Human CCL8 - Moderate). For research use only. Made in the USA
Catalog No. 50-289-2013 Supplier Kingfisher Biotech Inc Supplier No. KP2215S100
The Swine CCL8 polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Biotech Swine CCL8 ELISA. The Swine CCL8 antibody was produced in rabbits and is antigen-affinity purified. The reactivity by species is: (Swine CCL8 - Strong) (Bat CCL8 - None) (Bovine CCL8 - Moderate) (Canine CCL8 - None) (Cynomolgus Monkey CCL8 - None) (Equine CCL8 - None) (Feline CCL8 - None) (Ferret CCL8 - None) (Human CCL8 - Moderate). For research use only. Made in the USA