Learn More
Kingfisher Biotech Inc Human IL-6 ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY0722S003
Encompass_Preferred
The Human IL-6 ELISA contains capture antibody, standard, and detection antibody for development of a Human IL-6 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. A working knowledge of ELISA is strongly recommended. The Human IL-6 ELISA can also be used to measure Orangutan and Chimpanzee IL-6. Alternate names include IL6, BSF2, HGF, HSF, IFNB2, IL-6, BSF-2, CDF, IFN-beta-2, interleukin 6. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5225