Learn More
Kingfisher Biotech Inc Human TNF alpha ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1006S003
Encompass_Preferred
The Human TNF alpha (TNFSF2) ELISA contains capture antibody, protein standard, and detection antibody for development of a Human TNF alpha ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Human TNF alpha ELISA can also be used to measure Chimpanzee TNF alpha. Alternate names include TNF, DIF, TNF-alpha, TNFA, TNFSF2, Tumor necrosis factor, TNF-alpha, tumor necrosis factor, TNLG1F, Tumor necrosis factor alpha. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5309