Learn More
Med Vet International Swine Extra-Long 8mm Silicon Endotracheal Tube, J0615X
Catalog No. 5000581321
Encompass_Preferred
Especially designed with the cooperation of leading research centers for use in swine. Silicone. Graduated marking with standard adaptor, inflatable cuff, radio-paque, and lateral Murphy eye. Extra-long length. 8mmi.d. x 18"(47cm).
Catalog No. 50-005-81321 Supplier Med Vet International Supplier No. JOR615X
Especially designed with the cooperation of leading research centers for use in swine. Silicone. Graduated marking with standard adaptor, inflatable cuff, radio-paque, and lateral Murphy eye. Extra-long length. 8mmi.d. x 18"(47cm).