Learn More
Kingfisher Biotech Inc Human CXCL10 ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1149S004
Encompass_Preferred
The Human CXCL10 (IP-10) ELISA contains capture antibody, standard, and detection antibody for development of a Human CXCL10 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Human CXCL10 ELISA can also be used to measure Gorilla, Gibbon, and Chimpanzee CXCL10. Alternate names include CXCL10, C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1, C-X-C motif chemokine ligand 10, C-X-C motif chemokine 10. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5344