Learn More
Kingfisher Biotech Inc Ferret IL-1RA ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1947S004
Encompass_Preferred
The Ferret IL-1RA ELISA contains capture antibody, standard, and detection antibody for development of a Ferret IL-1RA ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. A working knowledge of ELISA is strongly recommended. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5614