Learn More
Kingfisher Biotech Inc Swine IL-10 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0301S025
Encompass
The Swine IL-10 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine IL-10 applications are for cell culture, ELISA standard, and Western Blot Control. The Swine IL-10 yeast-derived recombinant protein can be purchased in multiple sizes. Swine IL-10 Specifications: (Molecular Weight: 18.1 kDa) (Amino Acid Sequence: SIKSENSCIH FPTSLPHMLR ELRAAFGPVK SFFQTKDQMG DLLLTGSLLE DFKGYLGCQA LSEMIQFYLE DVMPKAESDG EDIKEHVNSL GEKLKTLRLR LRRCHQFLPC ENKSKAVEEV KSAFSKLQER GVYKAMGEFD IFINYIEAYM TMKMRKN (157)) (Gene ID: 397106). For research use only. Made in the USA
Catalog No. 50-241-3979