Learn More
Kingfisher Biotech Inc Swine IL-13 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0007S025
Encompass
The Swine IL-13 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine IL-13 applications are for cell culture, ELISA standard, and Western Blot Control. The Swine IL-13 yeast-derived recombinant protein can be purchased in multiple sizes. Swine IL-13 Specifications: (Molecular Weight: 13.2 kDa) (Amino Acid Sequence: LTCFGGLASP GPVPPHSTAL KELIEELVNI TQNQKTPLCN GSMVWSVNLT TSMYCAALES LINISDCSAI QKTQRMLSAL CSHKPPSEQV PGKHIRDTKI EVAQFVKDLL KHLRMIFRHG (120)) (Gene ID: 396721). For research use only. Made in the USA
Catalog No. 50-241-3862