Learn More
Kingfisher Biotech Inc Swine IL-8 ELISpot - Up to two (2) ELISpot assays
SDP
Catalog No. 502891848
Encompass_Preferred
Swine IL-8 ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-289-1848 Supplier Kingfisher Biotech Inc Supplier No. SPOT2554S004
Swine IL-8 ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA