Learn More
Kingfisher Biotech Inc Swine IL-22 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0262S025
Encompass
The Swine IL-22 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine IL-22 applications are for cell culture, ELISA standard, and Western Blot Control. The Swine IL-22 yeast-derived recombinant protein can be purchased in multiple sizes. Swine IL-22 Specifications: (Molecular Weight: 18.0 kDa) (Amino Acid Sequence: VPITHHCKLD QSNFQQPYIT NRTFTLAQEA SLADNNTDVR LIGNNLFQGV NMRERCYLVK QVLNFTLEEV LFPNSDRFHP YMQEVASFLD SLSKKLSQCR IKGDDQHIQR NVNNFKDTVK KLGESGEIKV IGELYLLFMA LKNECTLPGH SWKMDN (156)) (Gene ID: 595104). For research use only. Made in the USA
Catalog No. 50-241-3215