Learn More
Invitrogen™ Swine Influenza H1N1 HA Synthetic Peptide
Description
PEP-0829 can be used as a blocking peptide with polyclonal antibody PA5-20714. This peptide is recommended for use as a positive control in ELISA applications.
Specifications
Specifications
| Species | Virus |
| Purification Method | purified |
| Common Name | Swine Influenza H1N1 HA |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Gene Alias | flu hemagglutinin; H1N1 HA; H1N1 hemagglutinin; influenza a; Influenza H1N1; influenza hemagglutinin; swine H1N1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.