Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Swine Influenza H1N1 NS1 Synthetic Peptide

Catalog No. PEP0908 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0908 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0908 Supplier Invitrogen™ Supplier No. PEP0908
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide is recommended for use as a positive control in ELISA applications.

Influenza A H1N1 NS, also known as Non-structural protein 1 or NS1A or NS1, is a 234 amino acid containing protein belonging to the Influenza A virus (strain A/Swine/Iowa/15/1930 H1N1) which has aves, human and pig as host. This protein belongs to the influenza A viruses NS1 family and is a homodimer located in the nucleus of uninfected, transfected cells, and in the cytoplasm of infected cells. The viral protein NS1 binds to and inhibits two cellular proteins that are required for the 3'-end processing of cellular pre-mRNAs which include the 30 kDa cleavage and polyadenylation specificity factor (CPSF4) and the poly(A)-binding protein 2 (PABPN1). Thus inhibiting post-transcriptional processing of cellular pre-mRNA and leading to accumulation of unprocessed 3' end pre-mRNAs in nucleus and loss of cellular protein synthesis. This helps the virus to escape interferon synthesis after activation of the IRF3 pathway, and therefore prevents establishment of cellular antiviral state. NS1 is also known to prevent human EIF2AK2/PKR activation, at the very beginning of the infection, when NS1 is mainly present in the cytoplasm and prevent antiviral response.
TRUSTED_SUSTAINABILITY

Specifications

Species Virus
Purification Method purified
Common Name Swine Influenza H1N1 NS1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias Influenza NS1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.