Learn More
Kingfisher Biotech Inc Human IFN alpha ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY0751S004
Encompass_Preferred
The Human IFN alpha ELISA contains capture antibody, standard, and detection antibody for development of a Human IFN alpha ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA.
Catalog No. 50-228-5262