Learn More
Enterprise Technology Solutions Swingline Thermal Pouch Laminator, 9"W, White
SDP
Catalog No. 5000740121
Encompass_Preferred
Thermal pouch laminator warms up in as little as 4 minutes to quickly laminate your favorite photos, award certificates and other mementos. Single power setting eliminates the need to switch between settings based on pouch thickness for simple operation. Versatile design is compatible with 3 and 5 mil lamination pouches to provide durable protection that remains flexible. Cold option works with pressure-sensitive, self-adhesive lamination pouches. Jam-release lever easily clears misfed, wrapped or jammed pouches. Laminator includes one starter pack of letter-size, thermal lamination pouches.
Catalog No. 50-007-40121 Supplier Enterprise Technology Solutions Supplier No. SWI1701857CM
Thermal pouch laminator warms up in as little as 4 minutes to quickly laminate your favorite photos, award certificates and other mementos. Single power setting eliminates the need to switch between settings based on pouch thickness for simple operation. Versatile design is compatible with 3 and 5 mil lamination pouches to provide durable protection that remains flexible. Cold option works with pressure-sensitive, self-adhesive lamination pouches. Jam-release lever easily clears misfed, wrapped or jammed pouches. Laminator includes one starter pack of letter-size, thermal lamination pouches.