Learn More
StatLab Swingsette Embedding Cassettes
SDP
Catalog No. 23640198
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
23-640-198 Yellow
1 options
This item is not returnable. View return policy
Catalog No. 23-640-198 Supplier StatLab Supplier No. M5175SL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Most popular hinged cassette design
  • 45° labeling surface
  • Sleeved, hinged, front tab, yellow
  • Separate lids
Specifications
Quantity 1 Ea.
Color Yellow
Product Type Embedding Cassette