Learn More
Abcam Swinholide A, F-actin inhibitor, 50UG
Catalog No. 502705843
Encompass_Preferred
MW 1389.9 Da, Purity >97%. Cell permeable, selective F-actin inhibitor. Sequesters and stabilizes actin dimers. Induces actin depolymerization and apoptosis. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5843 Supplier Abcam Supplier No. AB14436650UG
MW 1389.9 Da, Purity >97%. Cell permeable, selective F-actin inhibitor. Sequesters and stabilizes actin dimers. Induces actin depolymerization and apoptosis. .
The product is subject to the following: Abcam Restricted Use Statement