Learn More
Sterlitech Corporation Swinnex, polypropylene, 13mm Filter Holder (5/Pk)
SDP
Supplier:  Sterlitech Corporation 520100
Encompass_Preferred
The Swinnex® line of syringe filter holders are used to ultraclean or sterilize small volumes of liquids dispensed by syringe. Connect the Swinnex® 13 mm filter holder with tubing to pressurized reservoir or automatic filling machine for filtering larger volumes.
Make your own syringe filter! Swinnex® filter holders give you the flexibility to select the membrane type and diameter that are most appropriate for your application. For sterile filtration, autoclave your Swinnex® filter holders with the membrane in place. Ultraclean or sterilize small volumes of liquids dispensed by a syringe or install in-line to filter larger volumes.
Catalog No. NC1681611