Learn More
McMaster-Carr High-Precision Tweezers
Catalog No. NC1457106
Encompass
High-Precision Tweezers 0.008" Wide x 0.008" Thick Standard Pointed Tips, Steel
Catalog No. NC1457106 Supplier McMaster-Carr Supplier No. 5184A43
High-Precision Tweezers 0.008" Wide x 0.008" Thick Standard Pointed Tips, Steel