Learn More
MSC Swiss Precision Instrument Inc. Thread Plug Gage, #10-32, Class 2B/3B, Go, Steel
Catalog No. 5000535943
Encompass
This thread plug gage is designed for inspecting internal threads, ensuring dimensional accuracy and compliance with specifications. Constructed from durable steel, it features a single-end design and is a 'Go' type gage, verifying that the thread is not undersized. It is suitable for #10-32 threads with 2B and 3B classifications.
  • Verifies internal thread accuracy
  • Constructed from durable steel
  • Single-end configuration for specific applications
  • Designed for #10-32 thread sizes
  • Compatible with 2B and 3B thread classifications
  • 'Go' gage type ensures threads meet minimum size requirements
  • NIST traceability certification for assured quality
  • Handle is not included
Catalog No. 50-005-35943 Supplier MSC Supplier No. 75887901
This thread plug gage is designed for inspecting internal threads, ensuring dimensional accuracy and compliance with specifications. Constructed from durable steel, it features a single-end design and is a 'Go' type gage, verifying that the thread is not undersized. It is suitable for #10-32 threads with 2B and 3B classifications.
  • Verifies internal thread accuracy
  • Constructed from durable steel
  • Single-end configuration for specific applications
  • Designed for #10-32 thread sizes
  • Compatible with 2B and 3B thread classifications
  • 'Go' gage type ensures threads meet minimum size requirements
  • NIST traceability certification for assured quality
  • Handle is not included