Learn More
Grainger Lutron Claro Rocker Switch, Single Pole, 120/277V AC, 15A, 1 ea
Catalog No. 502775633
Encompass_Preferred
The Claro 15 A single-pole rocker switch provides reliable, manual on/off control for indoor lighting or motor loads. Designed for a sleek, modern aesthetic, this maintained-action switch is engineered for simple in-wall installation and is compatible with a wide range of lighting types, including LED bulbs.
  • 15 A and 120/277 V AC electrical rating
  • Single-pole, maintained-action operation for one-location control
  • Large paddle design for intuitive, quiet switching
  • Gloss white finish fits standard decor
  • Compatible with LED, halogen, and incandescent load types
  • Screw terminals and push-in connections for versatile wiring
  • No neutral wire required for installation
Catalog No. 50-277-5633 Supplier Grainger Supplier No. 174V40
The Claro 15 A single-pole rocker switch provides reliable, manual on/off control for indoor lighting or motor loads. Designed for a sleek, modern aesthetic, this maintained-action switch is engineered for simple in-wall installation and is compatible with a wide range of lighting types, including LED bulbs.
  • 15 A and 120/277 V AC electrical rating
  • Single-pole, maintained-action operation for one-location control
  • Large paddle design for intuitive, quiet switching
  • Gloss white finish fits standard decor
  • Compatible with LED, halogen, and incandescent load types
  • Screw terminals and push-in connections for versatile wiring
  • No neutral wire required for installation