Learn More
Instech Five channel fluid swivel. 4 x microdialyisis channels (quartz-lined, .009in ID), 1 x 22ga channel. Power assisted for smooth rotation. For dual probe microdialysis.
SDP
Catalog No. 5081144
Encompass
The Instech MCS/5A is a five-channel stainless steel swivel designed for rats. To overcome the high rotational torque typically associated with multi-channel swivels, this model features a motorized, power-assist system that senses the animal's movement and drives the swivel core to follow it, minimizing stress on the subject.
Features
  • Type: Motorized five-channel fluid swivel
  • Channel Configuration: Four microdialysis channels and one 22ga infusion channel
  • Construction: Stainless steel body with self-adjusting seals
  • Operation: Proportional motor control for fine, continuous movement; counter-balanced to minimize forces
  • Compatibility: Designed for use with MC015/10 microdialysis tubing connectors
  • Output: Analog motor drive signal available for rotational activity monitoring
Catalog No. 50-811-44 Supplier Instech Supplier No. MCS/5A
The Instech MCS/5A is a five-channel stainless steel swivel designed for rats. To overcome the high rotational torque typically associated with multi-channel swivels, this model features a motorized, power-assist system that senses the animal's movement and drives the swivel core to follow it, minimizing stress on the subject.
Features
  • Type: Motorized five-channel fluid swivel
  • Channel Configuration: Four microdialysis channels and one 22ga infusion channel
  • Construction: Stainless steel body with self-adjusting seals
  • Operation: Proportional motor control for fine, continuous movement; counter-balanced to minimize forces
  • Compatibility: Designed for use with MC015/10 microdialysis tubing connectors
  • Output: Analog motor drive signal available for rotational activity monitoring