Learn More
Justrite™ Swivel Caster Set for Gas Cylinder Carts
Catalog No. 19085877 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
Gas Cylinder Carts
Catalog No. For Use With (Equipment)
19-085-877 Gas Cylinder Carts
1 options
Catalog No. 19-085-877 Supplier Justrite™ Supplier No. 35408
  • $338.00
    $231.65 / Each of 1
    Save $106.35
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Justrite™ Swivel Caster Set includes two 4-inch swivel casters and two 4-inch fixed casters, both with HDPE wheels, steel yokes and frame, with flat washer, lock washer and nut.
Replacement swivel caster set for gas-cylinder carts, includes two 4-inch swivel casters and two 4-inch fixed casters, both with HDPE wheels, steel yokes and frame, with flat washer, lock washer and nut. For gas cylinder cart models 35056 and 35058.
Compliance
OSHA, CGA P1, NFPA55
Warranty and Services
1 YEAR
Specifications
Type Swivel Caster Set
Material HDPE
Finish Powder-coated Paint
For Use With (Equipment) Gas Cylinder Carts
Weight (English) 6 lb.
Width (English) 4 in.