Learn More
BINDER™ Swivel Casters
SDP
Catalog No. p-200104336 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB 260, KB 470, KB 720, KB 1060
KB PRO Series
KBF PRO Series
KBF Series
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5738 KB 260, KB 470, KB 720, KB 1060
50-287-5622 KB PRO Series
50-287-5740 KBF Series
50-287-5741 KBF PRO Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-287-5738 Supplier BINDER™ Supplier No. 80122541
Only null left
Add to Cart
Add to Cart
Height-adjustable swivel casters
Height-adjustable swivel casters for horizontal alignment of unit on uneven floors.
Specifications
For Use With (Equipment) KB 260, KB 470, KB 720, KB 1060
Material Steel
Type Castor Frame