Learn More
Grainger Swivel Connector 1/2 CTS x 1/2 NPT
Catalog No. 502768349
Encompass_Preferred
The John Guest Female Swivel Connector is a plastic push-to-connect fitting that provides a leak-free connection for tubing in fluid transfer applications. It features a 360° rotating swivel for flexibility. The body is made of acetal copolymer plastic for chemical resistance. It has a pressure rating of 100 psi at 73°F. The fitting is compatible with 1/4" OD polyethylene tubing and has a 1/4" NPT female thread. It measures 0.63" in diameter and 0.94" in length. The connector requires no tools for installation.
Catalog No. 50-276-8349 Supplier Grainger Supplier No. 16T756
The John Guest Female Swivel Connector is a plastic push-to-connect fitting that provides a leak-free connection for tubing in fluid transfer applications. It features a 360° rotating swivel for flexibility. The body is made of acetal copolymer plastic for chemical resistance. It has a pressure rating of 100 psi at 73°F. The fitting is compatible with 1/4" OD polyethylene tubing and has a 1/4" NPT female thread. It measures 0.63" in diameter and 0.94" in length. The connector requires no tools for installation.