Learn More
MSC Vikan Floor Squeegee, Revolving Neck, 600 mm, Cellular Rubber, White
Catalog No. 502835559
Encompass
This revolving neck floor squeegee features a 600 mm cellular rubber blade, ideal for efficient removal of water and food debris. Its flexible blade adapts to surface contours, while the revolving neck accesses hard-to-reach areas. The easily removable cassette enhances cleanability and offers cost-effective blade replacement.
  • Designed for hygiene-critical environments.
  • Flexible blade efficiently removes water and debris.
  • Revolving neck reaches under and around equipment.
  • Removable cassette improves cleanability and reduces costs.
  • Durable construction, resistant to harsh chemicals.
  • Compatible with Euro threaded handles.
  • Complies with REACH, Halal, and Kosher standards.
  • Operates from -20 °C to 90 °C.
Catalog No. 50-283-5559 Supplier MSC Supplier No. 47166863
This revolving neck floor squeegee features a 600 mm cellular rubber blade, ideal for efficient removal of water and food debris. Its flexible blade adapts to surface contours, while the revolving neck accesses hard-to-reach areas. The easily removable cassette enhances cleanability and offers cost-effective blade replacement.
  • Designed for hygiene-critical environments.
  • Flexible blade efficiently removes water and debris.
  • Revolving neck reaches under and around equipment.
  • Removable cassette improves cleanability and reduces costs.
  • Durable construction, resistant to harsh chemicals.
  • Compatible with Euro threaded handles.
  • Complies with REACH, Halal, and Kosher standards.
  • Operates from -20 °C to 90 °C.