Learn More
MSC Vikan Floor Squeegee, Revolving Neck, 23.6 in, Yellow, 1 ea
Catalog No. 502869562
Encompass
This floor squeegee is designed for efficient removal of water and food debris from various flooring types. It features a double-bladed cellular rubber design for effective cleaning.
  • Revolving neck for optimal movement
  • Cassette is easily removable for cleaning or replacement
  • Yellow color for use in HACCP color-coded systems
  • Autoclave and dishwasher safe
Catalog No. 50-286-9562 Supplier MSC Supplier No. 47166855
This floor squeegee is designed for efficient removal of water and food debris from various flooring types. It features a double-bladed cellular rubber design for effective cleaning.
  • Revolving neck for optimal movement
  • Cassette is easily removable for cleaning or replacement
  • Yellow color for use in HACCP color-coded systems
  • Autoclave and dishwasher safe