Learn More
Instech Swivel holder for CMA/120 balance arms.
SDP
Catalog No. 5081057
Encompass
The Instech GIMBAL/CMA is a swivel holder designed for the CMA/120 balance arm, adding a second axis of movement to protect delicate microdialysis swivels. It prevents stress on the swivel by allowing multi‑axis motion and ensures stable operation during awake‑animal microdialysis or infusion procedures.
Features
  • Designed specifically for CMA/120 balance arms
  • Adds an extra axis of movement to reduce swivel stress
  • Prevents unintended swivel cap loosening during use
  • Recommended for use with Instech dual‑channel microdialysis swivels
  • Improves durability in long‑term freely moving animal studies
Catalog No. 50-810-57 Supplier Instech Supplier No. GIMBAL/CMA
The Instech GIMBAL/CMA is a swivel holder designed for the CMA/120 balance arm, adding a second axis of movement to protect delicate microdialysis swivels. It prevents stress on the swivel by allowing multi‑axis motion and ensures stable operation during awake‑animal microdialysis or infusion procedures.
Features
  • Designed specifically for CMA/120 balance arms
  • Adds an extra axis of movement to reduce swivel stress
  • Prevents unintended swivel cap loosening during use
  • Recommended for use with Instech dual‑channel microdialysis swivels
  • Improves durability in long‑term freely moving animal studies