Learn More
Grainger Starrett® Swivel Post Snug, For 659 and 675 Series, 1 ea.
Catalog No. 502786830
Encompass_Preferred
This swivel post snug is engineered for use with 659 and 675 series indicator holders. It provides robust support and versatility for precision inspection, layout, checking, and lineup operations, ensuring the indicator is securely positioned for accurate measurements.
  • Universally adjustable to any position
  • Enables indicator positioning at any height within the capacity of the upright base post
  • Allows 360° horizontal and vertical rotation
  • Features two 19 mm (3/4 in.) gripping holes
  • Designed for lengthwise indicator movement within the capacity of the 19 x 250 mm (3/4 x 9-7/8 in.) horizontal gage-holding rod
Catalog No. 50-278-6830 Supplier Grainger Supplier No. 197T20
This swivel post snug is engineered for use with 659 and 675 series indicator holders. It provides robust support and versatility for precision inspection, layout, checking, and lineup operations, ensuring the indicator is securely positioned for accurate measurements.
  • Universally adjustable to any position
  • Enables indicator positioning at any height within the capacity of the upright base post
  • Allows 360° horizontal and vertical rotation
  • Features two 19 mm (3/4 in.) gripping holes
  • Designed for lengthwise indicator movement within the capacity of the 19 x 250 mm (3/4 x 9-7/8 in.) horizontal gage-holding rod